Bottles, wide neck, with standard ground joints, WHEATON®
Leverancier: DWK Life Sciences
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
216017 216019 216020 216021 W216015
WHEA216017EA 692 EUR
WHEA216017 WHEA216019 WHEA216020 WHEA216021 WHEAW216015
Bottles, wide neck, with standard ground joints, WHEATON®
Flessen Glazen flessen
Borosilicate glass bottle and stopper.
  • Features no-drip pour lip
  • Ground glass stopper fits securely to prevent contamination
  • Easy to clean
  • Autoclavable
Wide mouth ideal for solids and viscous liquids.