Biopsiecassettes in QuickLoad™-kokers, Swingsette™, M518SL
Leverancier: Simport Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
M518-10SL M518-11SL M518-12SL M518-2SL M518-3SL M518-4SL M518-5SL M518-6SL M518-7SL M518-8SL M518-9SL
720-1485EA 179 EUR
720-1485 720-1486 720-1487 720-1491 720-1492 720-1493 720-1494 720-1495 720-1496 720-1497 720-1498
Biopsiecassettes in QuickLoad™-kokers, Swingsette™, M518SL
Histologie-cassettes Cassettes voor biopsiebehandeling
Acetal polymer.
  • Moulded from a special high density polymer, they keep specimens safely submerged in liquid
  • Totally resistant to the chemical action of histological solvents
  • The efficient flow-through slots maximise fluid exchange and ensure proper drainage
  • The special hinge make it easy to attach lid to base
Verpakking: Each case contains 10 sleeves of 75 cassettes and 10 bags of 75 lids.