Containers, met schroefdop
Leverancier: KARTELL
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
5623 5621 0563200 5633
KART5623EA 335 EUR
KART5623 KART5621 KART0563200 KART5633
Containers, met schroefdop
Diagnostische systemen Clinical Sample Collection/Preparation Faeces Sample Collection
Made of PS, with PE screw cap and spoon.
  • Disposable
  • Available sterile or non sterile
  • Good chemical resistance
  • Lightweight