Magnetische roerstaven, PTFE, ovaal
Leverancier: KARTELL
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
683 684 685 686 687 688 689
KART683EA 13.3 EUR
KART683 KART684 KART685 KART686 KART687 KART688 KART689
Magnetische roerstaven, PTFE, ovaal
Roerders Magnetische roerstaven
These oval shaped PTFE stirring bars are ideal for containers or bottles with round bottom.
  • Suitable for food stuff