Reageerbuisrekken van roestvrijstalen draad
Leverancier: BOCHEM
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
10170 10171 10172 10173 10174 10175 10176
BOCH10170EA 37.8 EUR
BOCH10170 BOCH10171 BOCH10172 BOCH10173 BOCH10174 BOCH10175 BOCH10176
Reageerbuisrekken van roestvrijstalen draad
Rekken Tube Racks
Test tube racks, made of stainless steel wire.
  • Very good temperature resistance
  • Good chemical resistance
  • Easy to clean