Heftafels voor laboratoria
Leverancier: BOCHEM
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
11015 11016 11020 11030 11040 11041 11080 11081 11090 11091 11031 11022 11032 11021
245-0029EA 240 EUR
245-0029 245-0030 245-0031 245-0034 245-0037 245-0038 245-0039 245-0040 245-0041 245-0042 245-0035 245-0033 245-0036 245-0032
Heftafels voor laboratoria
Heftafels voor laboratoria
Aluminium or anodised aluminium. These lab jacks feature an improved scissor technology with reduced tolerances. The scissor construction is made of 18/10 stainless steel.
  • Ergonomic knob, rounded and user friendly
  • Smaller models up to 240×240 mm have a simple adjusting wheel, which can be operated by hand
  • Bigger models from 300×300 mm are recommended to be used in combination with a ratchet, which connects with a special adjusting wheel (ratchet included)
Certificaten: DIN 12897