Nitrobenzeen 99%, extra pure
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
128420010 128420025 128420100
ACRO128420010EA 53.6 EUR
ACRO128420010 ACRO128420025 ACRO128420100
Nitrobenzeen 99%, extra pure
Nitrobenzeen
Formule: C₆H₅NO₂
Kookpunt: 210…211 °C (760 mmHg)
Smeltpunt: 5…6 °C
Dichtheid: 1,205 g/cm³
Vlampunt: 88 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00007043
CAS nummer: 98-95-3
EINECS: 202-716-0
UN: 1662
ADR: 6.1,II
Merck Index: 13,06621
Order Now
Specification Test Results
Appearance Clear yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5500 to 1.5540 (20°C, 589 nm)
Water ≤0.15 %
GC ≥98.5 %