Demountable cuvettes
Leverancier: HELLMA
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
106-0.50-40 106-0.10-40 106-0.20-40 106-0.01-40 124-0.1-40 124-0.01-40 124-0.2-40 124-0.5-40
634-9235EA 461 EUR
634-9235 634-9237 634-9239 HELL106-0.01-40 HELL124-0.1-40 HELL124-0.01-40 HELL124-0.2-40 HELL124-0.5-40
Demountable cuvettes
Cuvetten Cuvetten voor spectrofotometers
Demountable cuvettes for short pathlength being easier to clean used in microscopic examinations.
Suitable for high precision analytical techniques.
Bestelinformatie: A cell holder is usually required for use with photometers.