Inbed-ringen
Leverancier: KARTELL
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
2919 0291904 0291906 0291912 0291913
KART2919EA 81 EUR
KART2919 KART0291904 KART0291906 KART0291912 KART0291913
Inbed-ringen
Histologie-cassettes Inbedringen
Embedding rings made from ABS.
  • Dimensions: 41×28×11 mm
Available in the colours white, blue, yellow, green and pink.