Gaswasflessen, Drechsel-patroon, met GL-aansluiting
Leverancier: Lenz Laborglas GmbH & CO.KG
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
05500511 05500513 05510113 05510411 05500512 05500514 05500610 05510011 05510012 05510013 05510014 05510111 05510112 05510114 05510211 05510212 05510213 05510214 05510410 05510412
LENZ05500511EA 60.9 EUR
LENZ05500511 LENZ05500513 LENZ05510113 LENZ05510411 LENZ05500512 LENZ05500514 LENZ05500610 LENZ05510011 LENZ05510012 LENZ05510013 LENZ05510014 LENZ05510111 LENZ05510112 LENZ05510114 LENZ05510211 LENZ05510212 LENZ05510213 LENZ05510214 LENZ05510410 LENZ05510412
Gaswasflessen, Drechsel-patroon, met GL-aansluiting
Flessen Gaswasflessen
Borosilicate glass 3.3.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
With or without filter disc.
Certificaten: DIN 12596