Zonemonsternemer, Spiralus
Leverancier: BURKLE
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
5318-0145 5318-0200
BURK5318-0145EA 593 EUR
BURK5318-0145 BURK5318-0200
Zonemonsternemer, Spiralus
Monsternemers Monsternemers voor vaste stoffen
Guarantees precise zone sampling through all layers of the sample. The spiral arrangement of the sampling slits means that these are opened one after the other from bottom to top. In this way the lowest opening is filled first and then those above it in sequence until the complete Spiralus has been filled. The shape of the sampling slits prevents seeds from being cut up. The Spiralus provides an excellent all-layer sample.
  • Representative sample of the container layers.
  • Aluminium, anodised
  • Low weight
Spiralus is particularly suitable for sampling coarse-grained bulk goods, seeds and pellets at locations such as soil heaps, warehouses, trucks, railway goods wagons.