Media bottles, safety-coated, Lab 45™, WHEATON®
Leverancier: DWK Life Sciences
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
219940 219939 W219937
WHEA219940EA 536 EUR
WHEA219940 WHEA219939 WHEAW219937
Media bottles, safety-coated, Lab 45™, WHEATON®
Flessen Mediumflessen
Borosilicate glass, calr, Plastisol coated.
  • White 45 mm polypropylene screw cap attached
  • Pour ring eliminates drips
  • Autoclavable
  • Color coded caps available separately