Laboratory tables, with ground plate
Leverancier: BOCHEM
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
11324 11322 11321 11323 11310 11311 11312 11313 11314 11523 11524 11514 11522 11521 11510 11511 11512 11513 11924 11921 11783 11923 11782 11922 11784 11781
BOCH11324EA 6190 EUR
BOCH11324 BOCH11322 BOCH11321 BOCH11323 BOCH11310 BOCH11311 BOCH11312 BOCH11313 BOCH11314 BOCH11523 BOCH11524 BOCH11514 BOCH11522 BOCH11521 BOCH11510 BOCH11511 BOCH11512 BOCH11513 BOCH11924 BOCH11921 BOCH11783 BOCH11923 BOCH11782 BOCH11922 BOCH11784 BOCH11781
Laboratory tables, with ground plate
Meubilair Tafels
These laboratory tables are made up of 18/10 stainless steel and height adjustable.
  • Drawer face with user friendly handle bar
  • Noise protection
Working plate with matt finishing and corners are continuously welded and polished.
Drawer with guard rails made of stainless steel to mount GN-drawer. Drawers can be taken off for cleaning, but are secured against drawing out.