Apothekerspatel
Leverancier: RSG
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
651.150 651.130 651.100 651.075 051.150 051.130 051.100 051.075
RSGA651.150EA 59.1 EUR
RSGA651.150 RSGA651.130 RSGA651.100 RSGA651.075 RSGA051.150 RSGA051.130 RSGA051.100 RSGA051.075
Apothekerspatel
Spatels
Pharmacist's spatulas with stainless steel blade and black plastic handle.
  • Conical blade
  • Dishwasher-proof
  • Available flexible or non flexible