Kroestangen, met plastic handvat
Leverancier: BOCHEM
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
2162 2160 2161 2163
BOCH2162EA 15.6 EUR
BOCH2162 BOCH2160 BOCH2161 BOCH2163
Kroestangen, met plastic handvat
Tangen
Stainless steel crucible tongs with plastic handle (PVC).
  • Temperature resistant up to 145 °C
Order Now
Specificaties
Environmentally Preferable