Reageerbuisstatieven, Z-vormig
Leverancier: BOCHEM
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
8292 8293 8291 8290
212-0297EA 23.8 EUR
212-0297 212-0298 212-0296 212-0295
Reageerbuisstatieven, Z-vormig
Rekken Tube Racks
Racks, made of 18/10 stainless steel.
  • Very good temperature resistance
  • Good chemical resistance
  • Easy to clean