Bodemmonsternemer, GeoSampler
Leverancier: BURKLE
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
5350-5003 5350-5006
BURK5350-5003EA 121 EUR
BURK5350-5003 BURK5350-5006
Bodemmonsternemer, GeoSampler
Monsternemers Monsternemers voor vaste stoffen
GeoSampler is specially designed for soil sampling.
  • Depth graduation 10 cm divisions
  • With or without foot bar for easier work
Sampler for light soils, meadows and lawns.