Trays
Leverancier: BOCHEM
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
8735 8736
BOCH8735EA 88.2 EUR
BOCH8735 BOCH8736
Trays
Reservoirs/Schalen/Potten Laboratoriumbakken/-schalen
Tray made of stainless steel with two handles.
  • Very good temperature resistance
  • Very good chemical resistance
  • Easy to clean
Multipurpose trays for a wide range of uses.