Evaporating dishes, flat shape
Leverancier: BOCHEM
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
8571 8570 8572
BOCH8571EA 34.9 EUR
BOCH8571 BOCH8570 BOCH8572
Evaporating dishes, flat shape
Reservoirs/Schalen/Potten Uitdampschalen
Evaporating dishes are made of stainless steel 18/10.
  • Very good chemical resistance
  • Very good temperature resistance
  • Easy to clean