Uitdampschaaltjes
Leverancier: BOCHEM
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
8042 8045 8040 8041
BOCH8042EA 88.4 EUR
BOCH8042 BOCH8045 BOCH8040 BOCH8041
Uitdampschaaltjes
Reservoirs/Schalen/Potten Uitdampschalen
Made of nickel 99,5 %.
  • Good chemical resistance
  • Very good temperatrure resistance
  • Easy to clean
Without spout.