Blood collection tubes, with ACD, BD Vacutainer®
Leverancier: BD Medical
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
367756 366645
613-3989EA 93.8 EUR
613-3989 BDAM366645
Blood collection tubes, with ACD, BD Vacutainer®
Diagnostische systemen Clinical Sample Collection/Preparation Blood Sample Collection/Preparation
These ACD tubes are used to obtain a whole blood or a plasma sample.
  • Contains the active anticoagulant tri-sodium citrate with citric acid and dextrose
  • Available in two formulations, ACD solution A and ACD solution B
Tubes are available in glass only and are identifiable by a yellow cap.