L(-)-Glutathione (oxidised form)

Leverancier: Roth Carl
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synoniemen: L(-)-Glutathione, oxidized, Bi(glutathion-S-yl), GSSG, Glutathiol, Oxidized Glutathione (GSSG), Glutathione oxidized, (2S,2'S)-5,5'-(((2R,2'R)-Disulphanediylbis(1-((carboxymethyl)amino)-1-oxopropane-3,2-diyl))bis(azanediyl))bis(2-amino-5-oxopentanoic acid)

6378.1 6378.2
ROTH6378.1EA 0 EUR
ROTH6378.1 ROTH6378.2
L(-)-Glutathione (oxidised form)
L(-)-Glutathione (oxidised form)
Formule: C₂₀H₃₂N₆O₁₂S₂
MW: 612.64 g/mol
Smeltpunt: 185
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00063106
CAS nummer: 27025-41-8
EINECS: 248-170-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR