γ-Butyrolactone ≥99.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

Synoniemen: gamma-Butyrolactone, Butan-4-olide, 4-Hydroxybutanoic acid lactone, 1,4-Butyrolactone, GBL

B0767-500ML B0767-25ML
TCIAB0767-500MLEA 31.9 EUR
TCIAB0767-500ML TCIAB0767-25ML
γ-Butyrolactone ≥99.0% (by GC)
γ-Butyrolactone
Formule: C₄H₆O₂
MW: 86.09 g/mol
Kookpunt: 204
Smeltpunt: -44
Dichtheid: 1.1286
Vlampunt: 101°C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00005386
CAS nummer: 96-48-0
EINECS: 202-509-5
Merck Index: 14,01596
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=698f337a5e847b6fd31b841649b6f01e9dc6c1b4&vid=698f337a5e847b6fd31b841649b6f01e9dc6c1b4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 99.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR