ortho-Fosforzuur 75% ≥75%, EMPROVE® EXPERT for biopharmaceutical production, SAFC®
Leverancier: MERCK PRODUCTION CHEMICALS
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
Synoniemen: Phosphoric acid
1.00250.9010 1.00250.9950
1.00250.9010EA 350 EUR
1.00250.9010 1.00250.9950
ortho-Fosforzuur 75% ≥75%, EMPROVE® EXPERT for biopharmaceutical production, SAFC®
ortho-Fosforzuur
Formule: H₃PO₄
Kookpunt: 135 °C
Smeltpunt: –17 °C
Dichtheid: 1,58 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011340
CAS nummer: 7664-38-2
EINECS: 231-633-2
UN: 1805
ADR: 8,III
Order Now
Specification Test Results
Assay (alkalimetric) 74.0- 76.0 %
Residual solvents (ICH Q3C) ... excluded by production process
Endotoxins ≤ 2.5 I.U./ml