4-Bromobutyryl chloride 95%
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
303440050 303440250
ACRO303440050EA 44.4 EUR
ACRO303440050 ACRO303440250
4-Bromobutyryl chloride 95%
4-Bromobutyryl chloride
Formule: C₄H₆BrClO
Kookpunt: 101 °C (37 mmHg)
Dichtheid: 1,602 g/cm³
Vlampunt: 91 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00000753
CAS nummer: 927-58-2
EINECS: 213-154-0
UN: 3265
ADR: 8,II
Order Now
Specification Test Results
Appearance Clear colorless to light yellow to light pink Liquid
Infrared spectrum Conforms
Refractive index 1.4910 to 1.4940 (20°C, 589 nm)
GC ≥94 %