4-Chlorobutyryl chloride 98%
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
108902500 108900010
ACRO108902500EA 36.3 EUR
ACRO108902500 ACRO108900010
4-Chlorobutyryl chloride 98%
4-Chlorobutyryl chloride
Formule: C₄H₆Cl₂O
Kookpunt: 173…174 °C (760 mmHg)
Smeltpunt: –47 °C
Dichtheid: 1,25 g/cm³
Vlampunt: 85 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00000754
CAS nummer: 4635-59-0
EINECS: 225-059-1
UN: 2922
ADR: 8,I
Order Now
Specification Test Results
Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4605 to 1.4625 (20°C, 589 nm)
GC ≥99.0 %
Solubility (in water) Passes test
Color scale ≤10 APHA
Residue after evaporation ≤0.001 %
Carbonyl compounds ≤0.02 % 2-Butanone
Titrable acid ≤0.0005 meq/g