DL-allo-Threonine
Leverancier: BACHEM BIOCHEMICA
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
4016157.0250
4016157.1000
BACE4016157.0250EA
0
EUR
BACE4016157.0250
BACE4016157.1000
DL-allo-Threonine
DL-allo-Threonine
Formule:
C₄H₉NO₃ |
MDL Nummer:
MFCD00067248 CAS nummer: 144-98-9 EINECS: 205-645-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...