L-Threonine

Leverancier: Serva
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

36382.02 36382.03
SERA36382.02EA 0 EUR
SERA36382.02 SERA36382.03
L-Threonine
L-Threonine
Formule: C₄H₉NO₃
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00064270
CAS nummer: 72-19-5
EINECS: 200-774-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR