Isobutyrylchloride ≥98%
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
B24472.22
B24472.36
B24472.22EA
32.2
EUR
B24472.22
B24472.36
Isobutyrylchloride ≥98%
Isobutyrylchloride
Purity: 98%
CAS No.: 79-30-1
Molecular Formula: (CH3)2CHCOCl
Formula Weight: 106.55
Flash Point: 8°(46°F)
Refractive Index: 1.4070
Odour: Unpleasant
Melting Point: ca -90°
Boiling Point: 91-93°
Beilstein Reference: 605393
UN No.: 2395
MDL No.: MFCD00000717
Formule:
C₄H₇ClO Kookpunt: 91…93 °C Smeltpunt: ∼–90 °C Dichtheid: 1,017 Vlampunt: 8 °C (46 °F) Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00000717 CAS nummer: 79-30-1 EINECS: 201-194-1 UN: 2395 ADR: 3,II |