2,6-Dibromo-4-(trifluoromethoxy)aniline

Leverancier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

SPB05517.1 SPB05517.10
MAYBSPB05517.1EA 0 EUR
MAYBSPB05517.1 MAYBSPB05517.10
2,6-Dibromo-4-(trifluoromethoxy)aniline
2,6-Dibromo-4-(trifluoromethoxy)aniline
Formule: C₇H₄Br₂F₃NO
MW: 334.92 g/mol
MDL Nummer: MFCD00153113
CAS nummer: 88149-49-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR