2-(N-Ethylanilino)ethanol ≥98.0% (by GC, titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

Synoniemen: N-Ethyl-N-(2-hydroxyethyl)aniline, Ethylphenylethanolamine, 2-(Ethyl(phenyl)amino)ethanol, N-Ethyl-N-2-ethanolaniline

E0062-25G E0062-500G
TCIAE0062-25GEA 17.6 EUR
TCIAE0062-25G TCIAE0062-500G
2-(N-Ethylanilino)ethanol ≥98.0% (by GC, titration analysis)
2-(N-Ethylanilino)ethanol
Formule: C₁₀H₁₅NO
MW: 165.24 g/mol
MDL Nummer: MFCD00020575
CAS nummer: 92-50-2
EINECS: 202-160-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bcb5397e8cdcce19715c551459004399848562e1&vid=bcb5397e8cdcce19715c551459004399848562e1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Light yellow powder to lump
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR