2-Chloroanilinium chloride ≥97%
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
Synoniemen:
2-Chloroaniline hydrochloride
L13483.14
L13483.22
L13483.14EA
30
EUR
L13483.14
L13483.22
2-Chloroanilinium chloride ≥97%
2-Chloroanilinium chloride
Purity: 97%
CAS No.: 137-04-2
Molecular Formula: C6H6ClN.HCl
Formula Weight: 164.03
Odour: Characteristic
Melting Point: ca 220° subl.
Beilstein Reference: 3624280
UN No.: 2018
MDL No.: MFCD00053282
Formule:
C₆H₇Cl₂N MW: 164.03 g/mol Smeltpunt: ∼ 220 °C (sublimatie) Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00053282 CAS nummer: 137-04-2 EINECS: 205-274-7 UN: 2018 ADR: 6.1 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...