3-Chloroanilinium chloride

Leverancier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synoniemen: 3-Chloroaniline hydrochloride

SB01765.1 SB01765.10 SB01765.50
MAYBSB01765.1EA 0 EUR
MAYBSB01765.1 MAYBSB01765.10 MAYBSB01765.50
3-Chloroanilinium chloride
3-Chloroanilinium chloride
Formule: C₆H₇Cl₂N
MW: 164.03 g/mol
MDL Nummer: MFCD00035485
CAS nummer: 141-85-5
EINECS: 205-506-7
UN: 2018
ADR: 6.1 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR