Mycophenolic acid

Leverancier: Serva
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

29860.01 29860.02
SERA29860.01EA 0 EUR
SERA29860.01 SERA29860.02
Mycophenolic acid
Mycophenolic acid
Formule: C₁₇H₂₀O₆
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00036814
CAS nummer: 24280-93-1
EINECS: 246-119-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR