Sulphamide ≥99%
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
Synoniemen: Sulphuric diamide
A18680.09 A18680.18 A18680.88
A18680.09EA 82.3 EUR
A18680.09 A18680.18 A18680.88
Sulphamide ≥99%
Sulphamide
Purity: 99% CAS No.: 7803-58-9 Molecular Formula: SO2(NH2)2 Formula Weight: 96.11 Melting Point: 91-92° Solubility: Soluble in water, hot alcohol, acetone. Sparingly soluble in cold alcohol MDL No.: MFCD00011606
Formule: H₄N₂O₂S
MW: 96.11 g/mol
Smeltpunt: 91…92 °C
Dichtheid: 1.61
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011606
CAS nummer: 7803-58-9
EINECS: 232-262-9