Geraniol ≥97%
Leverancier: Thermo Fisher ScientificTotal Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
Purity: 97%
CAS No.: 106-24-1
Molecular Formula: C10H18O
Formula Weight: 154.25
Flash Point: 101°(213°F)
Refractive Index: 1.4760
Odour: Floral
Melting Point: -15°
Boiling Point: 114-115°/12mm
Beilstein Reference: 1722456
Merck Reference: 14,4403
MDL No.: MFCD00002917
Formule:
C₁₀H₁₈O Kookpunt: 114…115 °C (12 mmHg) Smeltpunt: –15 °C Dichtheid: 0,884 Vlampunt: 101 °C (213 °F) Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00002917 CAS nummer: 106-24-1 EINECS: 203-377-1 Merck Index: 14,4403 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...