2-(4-Fluorophenyl)indole
Leverancier: Thermo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
SEW02763.1
SEW02763.10
MAYBSEW02763.1EA
0
EUR
MAYBSEW02763.1
MAYBSEW02763.10
2-(4-Fluorophenyl)indole
2-(4-Fluorophenyl)indole
Formule:
C₁₄H₁₀FN MW: 211.24 g/mol |
MDL Nummer:
MFCD00068178 CAS nummer: 782-17-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...