3-Isocyanatomethyl-3,5,5-trimethylcyclohexylisocyanaat 98%
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
L13759.06
L13759.36
L13759.22
L13759.06EA
28.3
EUR
L13759.06
L13759.36
L13759.22
3-Isocyanatomethyl-3,5,5-trimethylcyclohexylisocyanaat 98%
3-Isocyanatomethyl-3,5,5-trimethylcyclohexylisocyanaat
Purity: 98%
CAS No.: 4098-71-9
Molecular Formula: C12H18N2O2
Formula Weight: 222.29
Storage Temperature: 2°C to 8°C
Flash Point: 163°(325°F)
Refractive Index: 1.4840
Melting Point: -60°
Boiling Point: 153°/10mm
Beilstein Reference: 2726467
UN No.: 2290
MDL No.: MFCD00064956
Formule:
C₁₂H₁₈N₂O₂ Kookpunt: 153 °C (10 mmHg) Smeltpunt: –60 °C Dichtheid: 1,061 Vlampunt: 163 °C (325 °F) |
MDL Nummer:
MFCD00064956 CAS nummer: 4098-71-9 EINECS: 223-861-6 UN: 2290 ADR: 6.1,III |