Diethyl bromomalonate 90+%
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
Synoniemen:
Diethyl 2-bromomalonate
114190250
114191000
114195000
ACRO114190250EA
51.7
EUR
ACRO114190250
ACRO114191000
ACRO114195000
Diethyl bromomalonate 90+%
Diethyl bromomalonate
Formule:
C₇H₁₁BrO₄ MW: 239.07 g/mol Kookpunt: 233…235 °C (760 mmHg) Dichtheid: 1.4 g/cm³ Vlampunt: >110 °C Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00009138 CAS nummer: 685-87-0 EINECS: 211-683-1 UN: 3265 ADR: 8 II |
Specification Test Results
Appearance | Clear colorless to light yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4510 to 1.4560 (20°C, 589 nm) |
GC | ≥90.0 % |
UV | at 214 nm A: ≤0.05 |
Titration with NaOH | ≥98.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...