Cyclopropylmethyl isothiocyanate

Leverancier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

S10467.250 S10467DA
MAYBS10467.250EA 0 EUR
MAYBS10467.250 MAYBS10467DA
Cyclopropylmethyl isothiocyanate
Cyclopropylmethyl isothiocyanate
Formule: C₅H₇NS
CAS nummer: 6068-90-2

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR