1-Propylboronic acid ≥98%
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
L19965.03
L19965.06
L19965.14
L19965.03EA
56.3
EUR
L19965.03
L19965.06
L19965.14
1-Propylboronic acid ≥98%
1-Propylboronic acid
Purity: 98%
CAS No.: 17745-45-8
Molecular Formula: CH3(CH2)2B(OH)2
Formula Weight: 87.91
Storage Temperature: 2°C to 8°C
Melting Point: 105-107°
Beilstein Reference: 1731884
MDL No.: MFCD01074564
Formule:
C₃H₉BO₂ Smeltpunt: 105…107 °C Bewaar temperatuur: Koelkast |
MDL Nummer:
MFCD01074564 CAS nummer: 17745-45-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...