Slangen, latex
Leverancier: KARTELL
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
3872 3870 3871 3873
KART3872EA 278 EUR
KART3872 KART3870 KART3871 KART3873
Slangen, latex
Slangen Transfer Tubing
Latex, translucent.
  • Density: 0,92
  • Autoclavable
Tubing with smooth and transparent surface.