Scalpels
Leverancier: BOCHEM
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
12640 12641 12642
BOCH12640EA 8.1 EUR
BOCH12640 BOCH12641 BOCH12642
Scalpels
Scalpels Scalpels
These mounted scalpels feature a steel blade and a coloured handle made of impact resistant PS.
  • Not sterile
  • Blades are exchangeable
  • Spare blades available (set of 10 pieces)
Order Now
Specificaties
Environmentally Preferable