Stoppen, diamant-geslepen
Leverancier: DWK Life Sciences
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
286121808 286122504 286123509 286124608 286126206 286129006 286130307 286130907 286127802
217-0796EA 290 EUR
217-0796 217-0797 217-0503 217-0800 217-0802 217-0806 217-0808 217-0809 217-0804
Stoppen, diamant-geslepen
Stoppen Glazen stoppen
DURAN®, borosilicate glass 3.3, clear.
  • Hexagonal
  • Flat
  • Diamond-ground