Stoppen, kort geslepen, DURAN®
Leverancier: DWK Life Sciences
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
286122607 286123809 286125004 286126609 286128001 286129409
217-0798EA 2.5 EUR
217-0798 217-0799 217-0801 217-0803 217-0805 217-0807
Stoppen, kort geslepen, DURAN®
Stoppen Glazen stoppen
DURAN®, borosilicate glass 3.3, clear.
  • Hexagonal
  • Short ground