Adapters, reductie, met conisch geslepen verbindingen, DURAN®
Leverancier: DWK Life Sciences
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
286542601 286543203 286548407 286548604 286549009 286549206 286549403 286524403 286524609 286524806 286536002 286536208
SCOT286542601EA 145 EUR
SCOT286542601 SCOT286543203 SCOT286548407 SCOT286548604 SCOT286549009 SCOT286549206 SCOT286549403 SCOT286524403 SCOT286524609 SCOT286524806 SCOT286536002 SCOT286536208
Adapters, reductie, met conisch geslepen verbindingen, DURAN®
Adapters Adapters met NS-verbindingen
DURAN®, borosilicate glass 3.3, clear.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Reduction adapter from large cone at the base to small socket on the top.