(Ethylthio)benzene ≥98%
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
Synoniemen: Ethyl phenyl sulphide
A13987.06 A13987.14 A13987.22
A13987.06EA 43.6 EUR
A13987.06 A13987.14 A13987.22
(Ethylthio)benzene ≥98%
(Ethylthio)benzene
Purity: 98% CAS No.: 622-38-8 Molecular Formula: C6H5SCH2CH3 Formula Weight: 138.23 Flash Point: 73°(163°F) Refractive Index: 1.5655 Odour: Stench Boiling Point: 204-205° Beilstein Reference: 1905731 MDL No.: MFCD00009265
Formule: C₈H₁₀S
MW: 138.23 g/mol
Kookpunt: 204…205 °C
Smeltpunt: 206.33
Dichtheid: 1.020
Vlampunt: 73 °C (163 °F)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00009265
CAS nummer: 622-38-8
EINECS: 210-731-9