Thiophenol 99%
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
138870010 138870250 138875000 138870050
ACRO138870010EA 159 EUR
ACRO138870010 ACRO138870250 ACRO138875000 ACRO138870050
Thiophenol 99%
Thiophenol
Formule: C₆H₆S
Kookpunt: 167…169 °C (760 mmHg)
Smeltpunt: –15 °C
Dichtheid: 1,078 g/cm³
Vlampunt: 50 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00004826
CAS nummer: 108-98-5
EINECS: 203-635-3
UN: 2337
ADR: 6.1,
Merck Index: 12,09492
Order Now
Specification Test Results
Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5880 to 1.5900 (20°C, 589 nm)
GC ≥98.5 %