Bowls
Leverancier: BOCHEM
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
8445 8446 8447 8448
216-0020EA 161 EUR
216-0020 216-0010 216-0021 BOCH8448
Bowls
Reservoirs/Schalen/Potten Laboratoriumbakken/-schalen
Made of stainless steel with two handles.
  • Very good chemical resistance
  • Very good thermal resistance
  • Smooth surfaces for easy cleaning
Multipurpose bowls ideal for a wide variety of uses.