Bunsen burners
Leverancier: BOCHEM
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
7000 7010
451-0179EA 116 EUR
451-0179 BOCH7010
Bunsen burners
Verwarmers Gasbranders
These gas burners have a maximum temperature of 1300 °C and are made up of anti-slip rubber coating under the base.
  • With air regulation
  • Pilot flame